missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfocalix[4]arene Hydrate 94.0+%, TCI America™

Catalog No. p-7135897
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04691G 1 g
S04695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04691G Supplier TCI America Supplier No. S04691G

Chemical Identifiers

CAS 112269-92-8
Molecular Formula C28H24O16S4
Molecular Weight (g/mol) 744.724
MDL Number MFCD03095486
InChI Key JFYBCAFLVNKHHG-UHFFFAOYSA-N
Synonym Tetrahydroxycalix[4]arenetetrasulfonic Acid, Tetrasulfo(tetrahydroxy)calix[4]arene
PubChem CID 644084
SMILES C1C2=CC(=CC(=C2O)CC3=CC(=CC(=C3O)CC4=C(C(=CC(=C4)S(=O)(=O)O)CC5=C(C1=CC(=C5)S(=O)(=O)O)O)O)S(=O)(=O)O)S(=O)(=O)O

Specifications

Quantity 1 g
Formula Weight 744.72
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfocalix[4]arene Hydrate