Learn More
Alkali Scientific 4"" Soft Rubber Roller
Catalog No. 502388786
Encompass
The 4" Soft Rubber Sealing Film Roller is a versatile and essential tool designed to facilitate the application of sealing films in laboratory settings. With its soft rubber construction, this roller ensures gentle yet effective pressure, allowing for smooth and uniform adhesion of sealing films to various labware, including microplates, PCR plates, and culture dishes. Its ergonomic design and comfortable grip make it easy to handle, minimizing the risk of fatigue during prolonged use. Whether sealing microplate wells for PCR, ELISA, or cell culture assays, the Soft Rubber Sealing Film Roller offers precise control and reliable performance, helping to maintain sample integrity and prevent contamination. Efficient and user-friendly, this roller streamlines laboratory workflows and contributes to the success of experimental procedures.
Catalog No. 502388786 Supplier Alkali Scientific Supplier No. SF4ML
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 4" Soft Rubber Sealing Film Roller is a versatile and essential tool designed to facilitate the application of sealing films in laboratory settings. With its soft rubber construction, this roller ensures gentle yet effective pressure, allowing for smooth and uniform adhesion of sealing films to various labware, including microplates, PCR plates, and culture dishes. Its ergonomic design and comfortable grip make it easy to handle, minimizing the risk of fatigue during prolonged use. Whether sealing microplate wells for PCR, ELISA, or cell culture assays, the Soft Rubber Sealing Film Roller offers precise control and reliable performance, helping to maintain sample integrity and prevent contamination. Efficient and user-friendly, this roller streamlines laboratory workflows and contributes to the success of experimental procedures.