missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Quinolinecarboxaldehyde 97.0+%, TCI America™
Supplier: TCI America Q006125G
Specifications
| 4-Quinolinecarboxaldehyde | |
| 50°C | |
| C10H7NO | |
| 25 g | |
| MGCGJBXTNWUHQE-UHFFFAOYSA-N | |
| quinoline-4-carbaldehyde | |
| 78072 | |
| ≥97.0% (GC,T) |
| 4363-93-3 | |
| 123°C | |
| MFCD00006781 | |
| 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde | |
| O=CC1=C2C=CC=CC2=NC=C1 | |
| 157.17 | |
| 157.17 | |
| Crystalline Powder |
Chemical Identifiers
| 4363-93-3 | |
| 157.17 | |
| MGCGJBXTNWUHQE-UHFFFAOYSA-N | |
| 78072 | |
| O=CC1=C2C=CC=CC2=NC=C1 |
| C10H7NO | |
| MFCD00006781 | |
| 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde | |
| quinoline-4-carbaldehyde |
Safety and Handling
TSCA : No