missing translation for 'onlineSavingsMsg'
Learn More

4-Quinolinecarboxaldehyde 97.0+%, TCI America™

Catalog No. Q00611G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00611G 1 g
Q00615G 5 g
Q006125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Q00611G Supplier TCI America Supplier No. Q00611G

Chemical Identifiers

CAS 4363-93-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00006781
InChI Key MGCGJBXTNWUHQE-UHFFFAOYSA-N
Synonym 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
PubChem CID 78072
IUPAC Name quinoline-4-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 50°C
Boiling Point 123°C
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Quinolinecarboxaldehyde