missing translation for 'onlineSavingsMsg'
Learn More

4-Quinolinecarboxaldehyde, 95%

CAS: 4363-93-3 | C10H7NO | 157.17 g/mol

$70.19 - $206.37

Chemical Identifiers

CAS 4363-93-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00006781
InChI Key MGCGJBXTNWUHQE-UHFFFAOYSA-N
Synonym 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
PubChem CID 78072
IUPAC Name quinoline-4-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC294820010
SDS View Documents
Thermo Scientific Chemicals
294820010
1 g Glass bottle
Each for $70.19
Only null left
 
AC294820050
SDS View Documents
Thermo Scientific Chemicals
294820050
5 g Glass bottle
Each for $206.37
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4363-93-3
157.17
MGCGJBXTNWUHQE-UHFFFAOYSA-N
78072
O=CC1=C2C=CC=CC2=NC=C1
C10H7NO
MFCD00006781
4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
quinoline-4-carbaldehyde

Specifications

4363-93-3
100.0
Brown-Yellow to White
Authentic
Glass bottle
MFCD00006781
4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
O=CC1=C2C=CC=CC2=NC=C1
157.17
157.17
Crystalline Powder, Chunks or Solid
94.0
45°C to 52°C
>112°C
94% min. (GC)
C10H7NO
1 g
MGCGJBXTNWUHQE-UHFFFAOYSA-N
quinoline-4-carbaldehyde
78072
95%
4-Quinolinecarboxaldehyde, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-453-0

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only