missing translation for 'onlineSavingsMsg'
Learn More

4-Quinolinecarboxaldehyde, 95%

Catalog No. AC294820010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294820010 1 g Glass bottle
AC294820050 5 g Glass bottle
AC294820250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC294820010 Supplier Thermo Scientific Chemicals Supplier No. 294820010

CAS: 4363-93-3 | C10H7NO | 157.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4363-93-3
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.17
MDL Number MFCD00006781
InChI Key MGCGJBXTNWUHQE-UHFFFAOYSA-N
Synonym 4-quinolinecarboxaldehyde, 4-quinolinecarbaldehyde, cinchoninaldehyde, quinoline-4-carboxaldehyde, 4-formylquinoline, unii-3a0k4yx051, chinolin-4-aldehyd, quinoline-4-aldehyde, 4quinolinecarboxaldehyde
PubChem CID 78072
IUPAC Name quinoline-4-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=NC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 45°C to 52°C
Color Brown-Yellow to White
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 157.17
Percent Purity 95%
Physical Form Crystalline Powder, Chunks or Solid
Chemical Name or Material 4-Quinolinecarboxaldehyde, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-453-0

RUO – Research Use Only