missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-5-ylbenzoic acid, ≥97%, Thermo Scientific™

Catalog No. CC44301CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC44301CB 250mg Amber Glass Bottle
CC44301DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC44301CB Supplier Thermo Scientific Chemicals Supplier No. CC44301CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 216959-91-0
Molecular Formula C11H8N2O2
Molecular Weight (g/mol) 200.197
MDL Number MFCD05864811
InChI Key CCBKADQVSXLSDN-UHFFFAOYSA-N
Synonym 4-pyrimidin-5-yl benzoic acid, 4-pyrimidin-5-yl-benzoic acid, 4-5-pyrimidinyl benzoic acid, benzoic acid, 4-5-pyrimidinyl
PubChem CID 4187247
IUPAC Name 4-pyrimidin-5-ylbenzoic acid
SMILES C1=CC(=CC=C1C2=CN=CN=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 4-Pyrimidin-5-ylbenzoic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.