missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrimidin-2-ylbenzoyl chloride, TECH, Thermo Scientific™

Chemical Identifiers

CAS 679806-84-9
Molecular Formula C11H7ClN2O
Molecular Weight (g/mol) 218.64
MDL Number MFCD09064974
InChI Key AQFRTZZTRGGBKO-UHFFFAOYSA-N
Synonym 4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride
PubChem CID 21092945
IUPAC Name 4-pyrimidin-2-ylbenzoyl chloride
SMILES C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC56202CB
SDS View Documents
Thermo Scientific Chemicals
CC56202CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

679806-84-9
218.64
AQFRTZZTRGGBKO-UHFFFAOYSA-N
21092945
C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl
C11H7ClN2O
MFCD09064974
4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride
4-pyrimidin-2-ylbenzoyl chloride

Specifications

679806-84-9
100.0
Amber Glass Bottle
MFCD09064974
4-pyrimidin-2-yl benzoyl chloride, benzoyl chloride,4-2-pyrimidinyl, 4-pyrimidin-2-yl-benzoyl chloride
C1=CN=C(N=C1)C2=CC=C(C=C2)C(=O)Cl
218.64
218.64
4-Pyrimidin-2-ylbenzoyl chloride
89.5
90%
C11H7ClN2O
250mg
AQFRTZZTRGGBKO-UHFFFAOYSA-N
4-pyrimidin-2-ylbenzoyl chloride
21092945
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents