missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyridylboronic Acid (contains varying amounts of Anhydride), TCI America™
$37.29 - $37.29
Chemical Identifiers
| CAS | 1692-15-5 |
|---|---|
| Molecular Formula | C5H6BNO2 |
| Molecular Weight (g/mol) | 122.92 |
| MDL Number | MFCD01074545 |
| InChI Key | QLULGIRFKAWHOJ-UHFFFAOYSA-N |
| PubChem CID | 2734379 |
| IUPAC Name | (pyridin-4-yl)boronic acid |
| SMILES | OB(O)C1=CC=NC=C1 |
Chemical Identifiers
| 1692-15-5 | |
| 122.92 | |
| QLULGIRFKAWHOJ-UHFFFAOYSA-N | |
| (pyridin-4-yl)boronic acid |
| C5H6BNO2 | |
| MFCD01074545 | |
| 2734379 | |
| OB(O)C1=CC=NC=C1 |
Specifications
| 1692-15-5 | |
| MFCD01074545 | |
| pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid | |
| OB(O)C1=CC=NC=C1 | |
| 122.92 | |
| 122.92 | |
| 4-Pyridylboronic Acid (contains varying amounts of Anhydride) |
| C5H6BNO2 | |
| 1 g | |
| QLULGIRFKAWHOJ-UHFFFAOYSA-N | |
| (pyridin-4-yl)boronic acid | |
| 2734379 | |
| Crystalline Powder |
Safety and Handling
TSCA : No