missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. P15941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P15941G Supplier TCI America Supplier No. P15941G

Chemical Identifiers

CAS 1692-15-5
Molecular Formula C5H6BNO2
Molecular Weight (g/mol) 122.92
MDL Number MFCD01074545
InChI Key QLULGIRFKAWHOJ-UHFFFAOYSA-N
PubChem CID 2734379
IUPAC Name (pyridin-4-yl)boronic acid
SMILES OB(O)C1=CC=NC=C1

Specifications

Quantity 1 g
Synonym pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid
Formula Weight 122.92
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridylboronic Acid (contains varying amounts of Anhydride)