missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinepropanol, Thermo Scientific Chemicals

Catalog No. AC440560050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC440560050 5 g
AC440560250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC440560050 Supplier Thermo Scientific Chemicals Supplier No. 440560050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2629-72-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
InChI Key PZVZGDBCMQBRMA-UHFFFAOYSA-N
IUPAC Name 3-(pyridin-4-yl)propan-1-ol
SMILES OCCCC1=CC=NC=C1

Specifications

Melting Point 35.0°C to 39.0°C
Boiling Point 289.0°C
Flash Point 113°C
Quantity 5 g
Formula Weight 137.81
Chemical Name or Material 4-Pyridinepropanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 220-107-8

RUO – Research Use Only