missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineethanethiol Hydrochloride 97.0+%, TCI America™

Catalog No. p-7135103
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13735G 5 g
P137325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13735G Supplier TCI America Supplier No. P13735G

Chemical Identifiers

CAS 6298-11-9
Molecular Formula C7H9ClNS
Molecular Weight (g/mol) 174.67
MDL Number MFCD00956047
InChI Key HXMNVHPGTRZNSP-UHFFFAOYSA-N
Synonym 4-(2-Mercaptoethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanethiol Hydrochloride
PubChem CID 11446637
IUPAC Name 2-pyridin-4-ylethanethiol;hydrochloride
SMILES C1=CN=CC=C1CCS.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 175.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridineethanethiol Hydrochloride