missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyridinecarboxaldehyde 96.0+%, TCI America™
$76.43 - $76.43
Chemical Identifiers
| CAS | 872-85-5 |
|---|---|
| Molecular Formula | C6H5NO |
| Molecular Weight (g/mol) | 107.112 |
| MDL Number | MFCD00006425 |
| InChI Key | BGUWFUQJCDRPTL-UHFFFAOYSA-N |
| Synonym | 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde |
| PubChem CID | 13389 |
| IUPAC Name | pyridine-4-carbaldehyde |
| SMILES | C1=CN=CC=C1C=O |
Chemical Identifiers
| 872-85-5 | |
| 107.112 | |
| BGUWFUQJCDRPTL-UHFFFAOYSA-N | |
| 13389 | |
| C1=CN=CC=C1C=O |
| C6H5NO | |
| MFCD00006425 | |
| 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde | |
| pyridine-4-carbaldehyde |
Specifications
| 872-85-5 | |
| C6H5NO | |
| 25 mL | |
| BGUWFUQJCDRPTL-UHFFFAOYSA-N | |
| pyridine-4-carbaldehyde | |
| 13389 | |
| ≥96.0% (GC) | |
| 4-Pyridinecarboxaldehyde |
| 187°C | |
| MFCD00006425 | |
| 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde | |
| C1=CN=CC=C1C=O | |
| 107.112 | |
| 107.11 | |
| Liquid |
Safety and Handling
RTECSNumber : NR9400000
TSCA : Yes