missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridinecarboxaldehyde 96.0+%, TCI America™

Catalog No. I014325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I014325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I014325ML Supplier TCI America Supplier No. I014325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 872-85-5
Molecular Formula C6H5NO
Molecular Weight (g/mol) 107.112
MDL Number MFCD00006425
InChI Key BGUWFUQJCDRPTL-UHFFFAOYSA-N
Synonym 4-pyridinecarboxaldehyde, isonicotinaldehyde, 4-formylpyridine, isonicotinic aldehyde, p-pyridinealdehyde, pyridine-4-carboxaldehyde, 4-pyridinealdehyde, 4-pyridylaldehyde, 4-pyridinecarbaldehyde, pyridine-4-aldehyde
PubChem CID 13389
IUPAC Name pyridine-4-carbaldehyde
SMILES C1=CN=CC=C1C=O

Specifications

Boiling Point 187°C
Quantity 25 mL
Formula Weight 107.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Pyridinecarboxaldehyde
RTECSNumber NR9400000
TSCA Yes