missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetonitrile hydrochloride, 98%

CAS: 92333-25-0 | C7H6N2 | 118.14 g/mol

$104.67 - $1147.45

Chemical Identifiers

CAS 92333-25-0
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00012825
InChI Key BMVSAKPRNWZCPG-UHFFFAOYSA-N
Synonym 4-pyridylacetonitrile hydrochloride, 2-pyridin-4-yl acetonitrile hydrochloride, 4-pyridyl acetonitrile hydrochloride, 4-pyridylacetonitrile hcl, 4-pyridineacetonitrile hydrochloride, pyridine-4-acetonitrile hydrochloride, 4-pyridineacetonitrile, monohydrochloride, 4-pyridineacetonitrilehcl, 4-cyanomethylpyridine hydrochloride, 4-pyridylaceto-nitrile hydrochloride
PubChem CID 12396608
SMILES N#CCC1=CC=NC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2509503
View Documents
Thermo Scientific Chemicals
B2509503
1 g
Each for $104.67
Only null left
 
AAB2509506
View Documents
Thermo Scientific Chemicals
B2509506
5 g
Each for $331.88
Only null left
 
AAB2509514
View Documents
Thermo Scientific Chemicals
B2509514
25 g
Each for $1,147.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

92333-25-0
118.14
BMVSAKPRNWZCPG-UHFFFAOYSA-N
12396608
C7H6N2
MFCD00012825
4-pyridylacetonitrile hydrochloride, 2-pyridin-4-yl acetonitrile hydrochloride, 4-pyridyl acetonitrile hydrochloride, 4-pyridylacetonitrile hcl, 4-pyridineacetonitrile hydrochloride, pyridine-4-acetonitrile hydrochloride, 4-pyridineacetonitrile, monohydrochloride, 4-pyridineacetonitrilehcl, 4-cyanomethylpyridine hydrochloride, 4-pyridylaceto-nitrile hydrochloride
N#CCC1=CC=NC=C1

Specifications

92333-25-0
C7H6N2
1 g
4-pyridylacetonitrile hydrochloride, 2-pyridin-4-yl acetonitrile hydrochloride, 4-pyridyl acetonitrile hydrochloride, 4-pyridylacetonitrile hcl, 4-pyridineacetonitrile hydrochloride, pyridine-4-acetonitrile hydrochloride, 4-pyridineacetonitrile, monohydrochloride, 4-pyridineacetonitrilehcl, 4-cyanomethylpyridine hydrochloride, 4-pyridylaceto-nitrile hydrochloride
N#CCC1=CC=NC=C1
118.14
154.6
4-Pyridineacetonitrile hydrochloride
∼260°C (decomposition)
MFCD00012825
UN3439
BMVSAKPRNWZCPG-UHFFFAOYSA-N
2-(pyridin-4-yl)acetonitrile
12396608
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only