missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetonitrile hydrochloride, 98%

Catalog No. AAB2509503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2509503 1 g
AAB2509506 5 g
AAB2509514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2509503 Supplier Thermo Scientific Chemicals Supplier No. B2509503

CAS: 92333-25-0 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92333-25-0
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD00012825
InChI Key BMVSAKPRNWZCPG-UHFFFAOYSA-N
Synonym 4-pyridylacetonitrile hydrochloride, 2-pyridin-4-yl acetonitrile hydrochloride, 4-pyridyl acetonitrile hydrochloride, 4-pyridylacetonitrile hcl, 4-pyridineacetonitrile hydrochloride, pyridine-4-acetonitrile hydrochloride, 4-pyridineacetonitrile, monohydrochloride, 4-pyridineacetonitrilehcl, 4-cyanomethylpyridine hydrochloride, 4-pyridylaceto-nitrile hydrochloride
PubChem CID 12396608
SMILES N#CCC1=CC=NC=C1

Specifications

Melting Point ∼260°C (decomposition)
Quantity 1 g
UN Number UN3439
IUPAC Name 2-(pyridin-4-yl)acetonitrile
Formula Weight 154.6
Percent Purity 98%
Chemical Name or Material 4-Pyridineacetonitrile hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only