missing translation for 'onlineSavingsMsg'
Learn More

4-pyridin-3-ylaniline, Thermo Scientific™

Catalog No. CC40014CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC40014CB 250mg Amber Glass Bottle
CC40014DA 1g Amber Glass Bottle
CC40014DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC40014CB Supplier Thermo Scientific Chemicals Supplier No. CC40014CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 82261-42-5
Molecular Formula C11H10N2
Molecular Weight (g/mol) 170.215
InChI Key DKFDPLVNPGJNDE-UHFFFAOYSA-N
Synonym 4-3-pyridyl aniline, 4-pyridin-3-yl aniline, 4-pyridin-3-yl-phenylamine, 4-3-pyridinyl aniline, 3-4-aminophenyl pyridine, benzenamine, 4-3-pyridinyl, 2-pyridyl-4-aniline, 4-pyridin-3-yl-phenylamine 2hcl salt, 4-3-pyridyl phenylamine, 3-p-aminophenyl pyridine
PubChem CID 459522
IUPAC Name 4-pyridin-3-ylaniline
SMILES C1=CC(=CN=C1)C2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 170.21
Percent Purity 95%
Chemical Name or Material 4-Pyridin-3-ylaniline