missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrid-4-ylbenzoic acid, 95%, Thermo Scientific™

Catalog No. p-3110720
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC35401CB 250 mg Amber Glass Bottle
CC35401DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC35401CB Supplier Thermo Scientific Chemicals Supplier No. CC35401CB
Only null left

Chemical Identifiers

CAS 4385-76-6
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD04114574
InChI Key DZLGZIGLHCRIMF-UHFFFAOYSA-N
Synonym 4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv
PubChem CID 1520811
SMILES OC(=O)C1=CC=C(C=C1)C1=CC=NC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-pyridin-4-ylbenzoic acid
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 4-Pyrid-4-ylbenzoic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.