missing translation for 'onlineSavingsMsg'
Learn More

[4-(Pyrid-2-yloxy)phenyl]methanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC48909DA

 View more versions of this product

Catalog No. CC48909DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

[4-(Pyrid-2-yloxy)phenyl]methanol
96.5
97%
C12H11NO2
1g
ZDTXOQRCIVCMLT-UHFFFAOYSA-N
[4-(pyridin-2-yloxy)phenyl]methanol
17859715
97%
194017-70-4
100.0
Amber Glass Bottle
MFCD08690278
4-pyrid-2-yloxy phenyl methanol, 4-pyridin-2-yloxy phenyl methanol, 4-2-pyridyloxy phenyl methanol, 4-pyridin-2-yl oxy phenyl, 2-4-hydroxymethylphenoxy pyridine, 4-2-pyridyloxy phenyl methan-1-ol, 4-pyridin-2-yl oxy phenyl methanol
OCC1=CC=C(OC2=CC=CC=N2)C=C1
201.23
201.22

Chemical Identifiers

194017-70-4
201.23
ZDTXOQRCIVCMLT-UHFFFAOYSA-N
17859715
OCC1=CC=C(OC2=CC=CC=N2)C=C1
C12H11NO2
MFCD08690278
4-pyrid-2-yloxy phenyl methanol, 4-pyridin-2-yloxy phenyl methanol, 4-2-pyridyloxy phenyl methanol, 4-pyridin-2-yl oxy phenyl, 2-4-hydroxymethylphenoxy pyridine, 4-2-pyridyloxy phenyl methan-1-ol, 4-pyridin-2-yl oxy phenyl methanol
[4-(pyridin-2-yloxy)phenyl]methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions