missing translation for 'onlineSavingsMsg'
Learn More

4-(pyrid-2-yloxy)benzaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC48904DA

Quantity 1g

 View more versions of this product

Catalog No. CC48904DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

194017-69-1
199.209
4-pyridin-2-yloxy benzaldehyde, 4-pyrid-2-yloxy benzaldehyde, 4-2-pyridyloxy benzaldehyde, benzaldehyde,4-2-pyridinyloxy, 4-2-pyridinyloxy benzaldehyde, 4-2-pyridinyloxy-benzaldehyde, 4-pyridin-2-yloxy-benzaldehyde, 4-2-pyridyloxy phenyl formaldehyde
4-pyridin-2-yloxybenzaldehyde
C12H9NO2
DPRZACGKYIDYCK-UHFFFAOYSA-N
3755292
C1=CC=NC(=C1)OC2=CC=C(C=C2)C=O

Specifications

4-(Pyrid-2-yloxy)benzaldehyde
97%
1g
DPRZACGKYIDYCK-UHFFFAOYSA-N
4-pyridin-2-yloxybenzaldehyde
3755292
97%
194017-69-1
C12H9NO2
4-pyridin-2-yloxy benzaldehyde, 4-pyrid-2-yloxy benzaldehyde, 4-2-pyridyloxy benzaldehyde, benzaldehyde,4-2-pyridinyloxy, 4-2-pyridinyloxy benzaldehyde, 4-2-pyridinyloxy-benzaldehyde, 4-pyridin-2-yloxy-benzaldehyde, 4-2-pyridyloxy phenyl formaldehyde
C1=CC=NC(=C1)OC2=CC=C(C=C2)C=O
199.209
199.21
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions