missing translation for 'onlineSavingsMsg'
Learn More

4-Pyrazolecarboxylic acid, 97%

Catalog No. AC396855000
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396855000 500 mg Glass bottle
AC396851000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC396855000 Supplier Thermo Scientific Chemicals Supplier No. 396855000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37718-11-9 | C4H4N2O2 | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37718-11-9
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00011558
InChI Key IMBBXSASDSZJSX-UHFFFAOYSA-N
Synonym 4-pyrazolecarboxylic acid, 4-carboxypyrazole, pyrazole-4-carboxylic acid, unii-4xjz84gv8e, 1h-pyrazole-4-carboxylicacid, 4-carboxy-1h-pyrazole, 4xjz84gv8e, pyrazole-4-carboxylic, pubchem13293, pubchem22749
PubChem CID 3015937
IUPAC Name 1H-pyrazole-4-carboxylic acid
SMILES C1=C(C=NN1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 282°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 112.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Pyrazolecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 253-641-5

RUO – Research Use Only