missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Propylcyclohexanone 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 98°C |
| Quantity | 5 g |
| Formula Weight | 140.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 4-Propylcyclohexanone |
Safety and Handling
| EINECSNumber | (3)-2411 |
| TSCA | No |