missing translation for 'onlineSavingsMsg'
Learn More

4-Propylbenzenesulfonyl Chloride 93.0+%, TCI America™

Catalog No. P13665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13665G 5 g
P136625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13665G Supplier TCI America Supplier No. P13665G
Only null left

Chemical Identifiers

CAS 146949-07-7
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.70
MDL Number MFCD00041508
InChI Key LEFGAGRZHLNPLS-UHFFFAOYSA-N
Synonym 4-n-propylbenzenesulfonyl chloride, 4-propylbenzene-1-sulfonyl chloride, 4-propyl-benzenesulfonylchloride, 4-propyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-propyl, p-propylbenzenesulfonyl chloride, 4-n-propylbenzenesulfonylchloride, acmc-209cxt, 4-propylphenylsulfonyl chloride, 4-propylphenylsulphonyl chloride
PubChem CID 2737222
IUPAC Name 4-propylbenzene-1-sulfonyl chloride
SMILES CCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 218.70
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Propylbenzenesulfonyl Chloride