missing translation for 'onlineSavingsMsg'
Learn More

4'-Propylacetophenone 95.0+%, TCI America™

Catalog No. p-7134914
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P103625G 25 g
P1036500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P103625G Supplier TCI America Supplier No. P103625G

Chemical Identifiers

CAS 2932-65-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00041359
InChI Key ZNBVIYMIVFKTIW-UHFFFAOYSA-N
Synonym p-propylacetophenone, 1-4-propylphenyl ethan-1-one, 1-4-propylphenyl ethanone, 4'-propylacetophenone, ethanone, 1-4-propylphenyl, 4-n-propylacetophenone, 4'-n-propylacetophenone, 1-acetyl-4-propylbenzene, p-propylacetophenon, p-n-propylacetophenone
PubChem CID 76236
IUPAC Name 1-(4-propylphenyl)ethan-1-one
SMILES CCCC1=CC=C(C=C1)C(C)=O

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Propylacetophenone
EINECSNumber (3)-3540
TSCA No