missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Propoxyacetophenone 98.0+%, TCI America™
Supplier: TCI America P23775G
Specifications
| 4′-Propoxyacetophenone | |
| 21°C | |
| 111°C | |
| MFCD01922144 | |
| RTYYKCQJSTZADZ-UHFFFAOYSA-N | |
| 1-(4-propoxyphenyl)ethanone | |
| 260132 | |
| ≥98.0% (GC) |
| 5736-86-7 | |
| White | |
| C11H14O2 | |
| 5 g | |
| CCCOC1=CC=C(C=C1)C(=O)C | |
| 178.231 | |
| 178.23 | |
| Crystalline Lumps |
Chemical Identifiers
| 5736-86-7 | |
| 178.231 | |
| RTYYKCQJSTZADZ-UHFFFAOYSA-N | |
| 1-(4-propoxyphenyl)ethanone |
| C11H14O2 | |
| MFCD01922144 | |
| 260132 | |
| CCCOC1=CC=C(C=C1)C(=O)C |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator