missing translation for 'onlineSavingsMsg'
Learn More

4'-Propoxyacetophenone 98.0+%, TCI America™

Catalog No. P23771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23771G 1 g
P23775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23771G Supplier TCI America Supplier No. P23771G

Chemical Identifiers

CAS 5736-86-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD01922144
InChI Key RTYYKCQJSTZADZ-UHFFFAOYSA-N
PubChem CID 260132
IUPAC Name 1-(4-propoxyphenyl)ethanone
SMILES CCCOC1=CC=C(C=C1)C(=O)C

Specifications

Melting Point 21°C
Color White
Boiling Point 111°C
Quantity 1 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Propoxyacetophenone
TSCA No
Recommended Storage Refrigerator