missing translation for 'onlineSavingsMsg'
Learn More

4-Propargylthiomorpholine 1,1-Dioxide 98.0+%, TCI America™

Catalog No. p-7135146
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14695G 5 g
P146925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14695G Supplier TCI America Supplier No. P14695G
Only null left

Chemical Identifiers

CAS 10442-03-2
Molecular Formula C7H11NO2S
Molecular Weight (g/mol) 173.23
MDL Number MFCD00085157
InChI Key CZBZIZOYSYHBNM-UHFFFAOYSA-N
PubChem CID 2777265
IUPAC Name 4-prop-2-ynyl-1,4-thiazinane 1,1-dioxide
SMILES C#CCN1CCS(=O)(=O)CC1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 5 g
Synonym 4-propargylthiomorpholine 1,1-dioxide, 4-prop-2-yn-1-yl thiomorpholine 1,1-dioxide, 4-2-propynyl thiomorpholine 1,1-dioxide, 4-propargyl thiomorpholine-1,1-dioxide, 4-propargylthiomorpholine1,1-dioxide, thiomorpholine, 4-2-propyn-1-yl-, 1,1-dioxide, 4-prop-2-yn-1-yl-1??-thiomorpholine-1,1-dione, acmc-1byom, 4-propargylthiomorpholine-1,1-dioxide, 4-propargyl-thiomorpholine-1,1-dioxide
Formula Weight 173.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Propargylthiomorpholine 1,1-Dioxide