missing translation for 'onlineSavingsMsg'
Learn More

4-(piperidinomethyl)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 159691-33-5
Molecular Formula C13H17NO2
Molecular Weight (g/mol) 219.284
InChI Key RNCCMTVMMFUIKR-UHFFFAOYSA-N
Synonym 4-piperidin-1-ylmethyl benzoic acid, 4-piperidin-1-ylmethyl-benzoic acid, benzoic acid,4-1-piperidinylmethyl, 4-piperidylmethyl benzoic acid, 4-piperidinomethyl benzoic acid, 4-piperidin-1-yl methyl benzoic acid
PubChem CID 1092975
IUPAC Name 4-(piperidin-1-ylmethyl)benzoic acid
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC45501CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45501DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45501EA
View Documents Promotion Details
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

159691-33-5
219.284
4-piperidin-1-ylmethyl benzoic acid, 4-piperidin-1-ylmethyl-benzoic acid, benzoic acid,4-1-piperidinylmethyl, 4-piperidylmethyl benzoic acid, 4-piperidinomethyl benzoic acid, 4-piperidin-1-yl methyl benzoic acid
4-(piperidin-1-ylmethyl)benzoic acid
C13H17NO2
RNCCMTVMMFUIKR-UHFFFAOYSA-N
1092975
C1CCN(CC1)CC2=CC=C(C=C2)C(=O)O

Specifications

159691-33-5
C13H17NO2
RNCCMTVMMFUIKR-UHFFFAOYSA-N
4-(piperidin-1-ylmethyl)benzoic acid
1092975
97%
97%
4-piperidin-1-ylmethyl benzoic acid, 4-piperidin-1-ylmethyl-benzoic acid, benzoic acid,4-1-piperidinylmethyl, 4-piperidylmethyl benzoic acid, 4-piperidinomethyl benzoic acid, 4-piperidin-1-yl methyl benzoic acid
C1CCN(CC1)CC2=CC=C(C=C2)C(=O)O
219.284
219.28
4-(Piperidinomethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents