missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinemethanol 97.0+%, TCI America™

Catalog No. p-7135192
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16535G 5 g
P165325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16535G Supplier TCI America Supplier No. P16535G

Chemical Identifiers

CAS 6457-49-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
InChI Key XBXHCBLBYQEYTI-UHFFFAOYSA-N
Synonym 4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol
PubChem CID 420771
IUPAC Name piperidin-4-ylmethanol
SMILES C1CNCCC1CO

Specifications

Melting Point 59°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 3263
Formula Weight 115.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Piperidinemethanol
EINECSNumber (5)-3729
TSCA No