missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinemethanol, 98%

Catalog No. p-3737230
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC380630250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC380630250 Supplier Thermo Scientific Chemicals Supplier No. 380630250

CAS: 6457-49-4 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6457-49-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
MDL Number MFCD00174228
InChI Key XBXHCBLBYQEYTI-UHFFFAOYSA-N
Synonym 4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol
PubChem CID 420771
IUPAC Name piperidin-4-ylmethanol
SMILES C1CNCCC1CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 57.0°C to 61.0°C
Color White
Boiling Point 118.0°C to 120.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 115.17
Percent Purity 98%
Physical Form Crystalline Powder, Chunks or Solid
Chemical Name or Material 4-Piperidinemethanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only