missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinemethanol, 97%

Catalog No. AAL1796414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1796414 25 g
AAL1796406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1796414 Supplier Thermo Scientific Chemicals Supplier No. L1796414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6457-49-4 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6457-49-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00174228
InChI Key XBXHCBLBYQEYTI-UHFFFAOYSA-N
Synonym 4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol
PubChem CID 420771
IUPAC Name piperidin-4-ylmethanol
SMILES C1CNCCC1CO

Specifications

Melting Point 57°C to 61°C
Density 0.997
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Quantity 25 g
UN Number UN3259
Beilstein 103071
Sensitivity Hygroscopic
Formula Weight 115.18
Percent Purity 97%
Chemical Name or Material 4-Piperidinemethanol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only