missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Piperidinecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America I0256500G
Specifications
| 4-Piperidinecarboxylic Acid | |
| 336°C | |
| MFCD00006004 | |
| isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie | |
| OC(=O)C1CCNCC1 | |
| 129.16 | |
| 129.16 | |
| Crystalline Powder |
| 498-94-2 | |
| C6H11NO2 | |
| 500 g | |
| SRJOCJYGOFTFLH-UHFFFAOYSA-N | |
| piperidine-4-carboxylic acid | |
| 3773 | |
| ≥98.0% (T) |
Chemical Identifiers
| 498-94-2 | |
| 129.16 | |
| SRJOCJYGOFTFLH-UHFFFAOYSA-N | |
| 3773 | |
| OC(=O)C1CCNCC1 |
| C6H11NO2 | |
| MFCD00006004 | |
| isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie | |
| piperidine-4-carboxylic acid |
Safety and Handling
EINECSNumber : (5)-0770
RTECSNumber : NS5150000
TSCA : Yes