missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. I025625G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025625G 25 g
I0256100G 100 g
I0256500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I025625G Supplier TCI America Supplier No. I025625G

Chemical Identifiers

CAS 498-94-2
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00006004
InChI Key SRJOCJYGOFTFLH-UHFFFAOYSA-N
Synonym isonipecotic acid, 4-piperidinecarboxylic acid, 4-carboxypiperidine, hexahydroisonicotinic acid, dl-isopipecolinic acid, isonicotinic acid, hexahydro, h-inp-oh, acide isonipecotique, 4-hexahydroisonicotinic acid, unii-m5tzp1rwie
PubChem CID 3773
IUPAC Name piperidine-4-carboxylic acid
SMILES OC(=O)C1CCNCC1

Specifications

Melting Point 336°C
Quantity 25 g
Formula Weight 129.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Piperidinecarboxylic Acid
EINECSNumber (5)-0770
RTECSNumber NS5150000
TSCA Yes