missing translation for 'onlineSavingsMsg'
Learn More

4-(Piperidin-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 727733-92-8
Molecular Formula C13H16N2
Molecular Weight (g/mol) 200.285
MDL Number MFCD06796182
InChI Key JOGVBKNLRQCYKS-UHFFFAOYSA-N
Synonym 4-piperidin-1-ylmethyl benzonitrile, 1-4-cyanobenzyl piperidine, 4-piperidinomethyl benzonitrile, 4-piperidylmethyl benzenecarbonitrile, 4-piperidin-1-ylmethyl-benzonitrile, benzonitrile,4-1-piperidinylmethyl
PubChem CID 7537603
IUPAC Name 4-(piperidin-1-ylmethyl)benzonitrile
SMILES C1CCN(CC1)CC2=CC=C(C=C2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CC45516DA
View Documents
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC45516DE
View Documents
N/A 5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

727733-92-8
200.285
JOGVBKNLRQCYKS-UHFFFAOYSA-N
7537603
C1CCN(CC1)CC2=CC=C(C=C2)C#N
C13H16N2
MFCD06796182
4-piperidin-1-ylmethyl benzonitrile, 1-4-cyanobenzyl piperidine, 4-piperidinomethyl benzonitrile, 4-piperidylmethyl benzenecarbonitrile, 4-piperidin-1-ylmethyl-benzonitrile, benzonitrile,4-1-piperidinylmethyl
4-(piperidin-1-ylmethyl)benzonitrile

Specifications

727733-92-8
100.0
Amber Glass Bottle
MFCD06796182
JOGVBKNLRQCYKS-UHFFFAOYSA-N
4-(piperidin-1-ylmethyl)benzonitrile
7537603
97%
96.5
97%
C13H16N2
4-piperidin-1-ylmethyl benzonitrile, 1-4-cyanobenzyl piperidine, 4-piperidinomethyl benzonitrile, 4-piperidylmethyl benzenecarbonitrile, 4-piperidin-1-ylmethyl-benzonitrile, benzonitrile,4-1-piperidinylmethyl
C1CCN(CC1)CC2=CC=C(C=C2)C#N
200.285
200.28
4-(Piperidin-1-ylmethyl)benzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents