missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline, 98%

Catalog No. p-3598272
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157602500 250 mL Glass bottle
AC157600010 1 L Glass bottle
AC157600025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157602500 Supplier Thermo Scientific Chemicals Supplier No. 157602500
Only null left

CAS: 108-89-4 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-89-4
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00006440
InChI Key FKNQCJSGGFJEIZ-UHFFFAOYSA-N
Synonym 4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
PubChem CID 7963
ChEBI CHEBI:32547
IUPAC Name 4-methylpyridine
SMILES CC1=CC=NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.4°C
Color Yellow
Density 0.9500g/mL
Boiling Point 145°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5040 to 1.506
Quantity 250 mL
Beilstein 20, 240
Specific Gravity 0.95
Merck Index 15, 7514
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 93.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Picoline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-626-4

RUO – Research Use Only