missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline, 98%

Catalog No. AAA15595AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15595AP 500 mL
AAA15595AE 100 mL
AAA155950F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15595AP Supplier Thermo Scientific Chemicals Supplier No. A15595AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-89-4 | C6H7N | 93.129 g/mol

4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-vinylpyridine is produced from 4-Picoline, also used to produce several catalysts like 4-DMAP.

Solubility
Soluble in water, alcohol and ether.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, water & moisture.

Chemical Identifiers

CAS 108-89-4
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006440
InChI Key FKNQCJSGGFJEIZ-UHFFFAOYSA-N
Synonym 4-picoline, pyridine, 4-methyl, p-picoline, gamma-picoline, p-methylpyridine, 4-methyl-pyridine, gamma-methylpyridine, unii-tjd6v9sso7, .gamma.-picoline, 4-mepy
PubChem CID 7963
ChEBI CHEBI:32547
IUPAC Name 4-methylpyridine
SMILES CC1=CC=NC=C1

Specifications

Melting Point 3°C
Density 0.957
pH 10 to 11
Boiling Point 144°C to 145°C
Flash Point 39°C (102°F)
Odor Unpleasant
Refractive Index 1.505
Quantity 500 mL
UN Number UN2313
Beilstein 104586
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,7402
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 93.13
Percent Purity 98%
Chemical Name or Material 4-Picoline
Show More Show Less
Hazard Category H226-H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H226-H302-H332-H315-H319-H335
Toxic in contact with skin.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PICOLINES
EINECSNumber 203-626-4
RTECSNumber UT5425000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only