missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-phenyltetrahydropyran-4-yl)methylamine, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 14006-32-7 |
|---|---|
| Molecular Formula | C12H17NO |
| Molecular Weight (g/mol) | 191.27 |
| MDL Number | MFCD02946795 |
| InChI Key | IPYDNTFWMDEOBW-UHFFFAOYSA-N |
| Synonym | 4-phenyloxan-4-yl methanamine, 4-phenyltetrahydro-2h-pyran-4-yl methanamine, 4-phenyltetrahydropyran-4-yl methylamine, 1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, c-4-phenyl-tetrahydro-pyran-4-yl-methylamine, 4-phenyltetrahydro-2h-pyran-4-yl methylamine, 1-4-phenyloxan-4-yl methanamine, 2h-pyran-4-methanamine, tetrahydro-4-phenyl, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine, 4-phenyl-tetrahydro-2h-pyran-4-methylamine |
| PubChem CID | 6483779 |
| IUPAC Name | 1-(4-phenyloxan-4-yl)methanamine |
| SMILES | NCC1(CCOCC1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC69113CB
|
Thermo Scientific Chemicals
CC69113CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC69113DA
|
Thermo Scientific Chemicals
CC69113DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC69113DE
|
Thermo Scientific Chemicals
CC69113DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 14006-32-7 | |
| 191.27 | |
| IPYDNTFWMDEOBW-UHFFFAOYSA-N | |
| 6483779 | |
| NCC1(CCOCC1)C1=CC=CC=C1 |
| C12H17NO | |
| MFCD02946795 | |
| 4-phenyloxan-4-yl methanamine, 4-phenyltetrahydro-2h-pyran-4-yl methanamine, 4-phenyltetrahydropyran-4-yl methylamine, 1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, c-4-phenyl-tetrahydro-pyran-4-yl-methylamine, 4-phenyltetrahydro-2h-pyran-4-yl methylamine, 1-4-phenyloxan-4-yl methanamine, 2h-pyran-4-methanamine, tetrahydro-4-phenyl, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine, 4-phenyl-tetrahydro-2h-pyran-4-methylamine | |
| 1-(4-phenyloxan-4-yl)methanamine |
Specifications
| 14006-32-7 | |
| Amber Glass Bottle | |
| MFCD02946795 | |
| 4-phenyloxan-4-yl methanamine, 4-phenyltetrahydro-2h-pyran-4-yl methanamine, 4-phenyltetrahydropyran-4-yl methylamine, 1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, c-4-phenyl-tetrahydro-pyran-4-yl-methylamine, 4-phenyltetrahydro-2h-pyran-4-yl methylamine, 1-4-phenyloxan-4-yl methanamine, 2h-pyran-4-methanamine, tetrahydro-4-phenyl, 4-phenyl-2h-3,4,5,6-tetrahydropyran-4-yl methylamine, 4-phenyl-tetrahydro-2h-pyran-4-methylamine | |
| NCC1(CCOCC1)C1=CC=CC=C1 | |
| 191.27 | |
| 191.27 | |
| (4-phenyltetrahydropyran-4-yl)methylamine |
| 95% | |
| C12H17NO | |
| 250mg | |
| IPYDNTFWMDEOBW-UHFFFAOYSA-N | |
| 1-(4-phenyloxan-4-yl)methanamine | |
| 6483779 | |
| 95% |
Safety and Handling
GHS Signal Word: Danger