missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™

$110.12 - $110.12

Chemical Identifiers

CAS 182491-21-0
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00085758
InChI Key BWHJLSRDMNLSET-UHFFFAOYSA-N
Synonym 4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
PubChem CID 4138553
IUPAC Name 4-phenyloxane-4-carboxylic acid
SMILES OC(=O)C1(CCOCC1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC69101CB
SDS View Documents
Thermo Scientific Chemicals
CC69101CB
250 mg Amber Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

182491-21-0
206.24
BWHJLSRDMNLSET-UHFFFAOYSA-N
4138553
OC(=O)C1(CCOCC1)C1=CC=CC=C1
C12H14O3
MFCD00085758
4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
4-phenyloxane-4-carboxylic acid

Specifications

182491-21-0
Amber Glass Bottle
MFCD00085758
4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
OC(=O)C1(CCOCC1)C1=CC=CC=C1
206.24
206.24
4-Phenyltetrahydropyran-4-carboxylic acid
97%
C12H14O3
250 mg
BWHJLSRDMNLSET-UHFFFAOYSA-N
4-phenyloxane-4-carboxylic acid
4138553
97%
Videos
Safety and Handling

Safety and Handling

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'