missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylpyridine 98.0+%, TCI America™
$100.67 - $287.06
Chemical Identifiers
| CAS | 939-23-1 |
|---|---|
| Molecular Formula | C11H9N |
| Molecular Weight (g/mol) | 155.20 |
| MDL Number | MFCD00006420 |
| InChI Key | JVZRCNQLWOELDU-UHFFFAOYSA-N |
| Synonym | pyridine, 4-phenyl, p-phenylpyridine, 4-phenyl-pyridine, 4-azabiphenyl, unii-452kd9ycg7, 4-aza-1,1'-biphenyl, phenyl-pyridine, 4-phenylpyridin, 4-pyridylbenzene, zlchem 480 |
| PubChem CID | 13651 |
| IUPAC Name | 4-phenylpyridine |
| SMILES | C1=CC=C(C=C1)C1=CC=NC=C1 |
Chemical Identifiers
| 939-23-1 | |
| 155.20 | |
| JVZRCNQLWOELDU-UHFFFAOYSA-N | |
| 13651 | |
| C1=CC=C(C=C1)C1=CC=NC=C1 |
| C11H9N | |
| MFCD00006420 | |
| pyridine, 4-phenyl, p-phenylpyridine, 4-phenyl-pyridine, 4-azabiphenyl, unii-452kd9ycg7, 4-aza-1,1'-biphenyl, phenyl-pyridine, 4-phenylpyridin, 4-pyridylbenzene, zlchem 480 | |
| 4-phenylpyridine |
Specifications
| 939-23-1 | |
| 281°C | |
| MFCD00006420 | |
| pyridine, 4-phenyl, p-phenylpyridine, 4-phenyl-pyridine, 4-azabiphenyl, unii-452kd9ycg7, 4-aza-1,1'-biphenyl, phenyl-pyridine, 4-phenylpyridin, 4-pyridylbenzene, zlchem 480 | |
| C1=CC=C(C=C1)C1=CC=NC=C1 | |
| 155.20 | |
| 155.20 | |
| Crystalline Powder |
| 75°C | |
| C11H9N | |
| 5 g | |
| JVZRCNQLWOELDU-UHFFFAOYSA-N | |
| 4-phenylpyridine | |
| 13651 | |
| ≥98.0% (GC) | |
| 4-Phenylpyridine |
Safety and Handling
EINECSNumber : (5)-3717
RTECSNumber : UT7141000
TSCA : Yes