missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Phenylpyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 75°C |
| Boiling Point | 281°C |
| Quantity | 25 g |
| Formula Weight | 155.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Phenylpyridine |
Safety and Handling
| EINECSNumber | (5)-3717 |
| RTECSNumber | UT7141000 |
| TSCA | Yes |