missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpiperidine 98.0+%, TCI America™

Catalog No. P15551G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P15551G Supplier TCI America Supplier No. P15551G

Chemical Identifiers

CAS 771-99-3
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00006002
InChI Key UTBULQCHEUWJNV-UHFFFAOYSA-N
Synonym 4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
PubChem CID 69873
IUPAC Name 4-phenylpiperidine
SMILES C1CNCCC1C2=CC=CC=C2

Specifications

Melting Point 63°C
Color White
Boiling Point 257°C
Quantity 1 g
Formula Weight 161.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylpiperidine