missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylpiperidine, 96%

Catalog No. p-7057060
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6638903 1 g
AAH6638906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6638903 Supplier Thermo Scientific Chemicals Supplier No. H6638903
Only null left

CAS: 771-99-3 | C11H15N | 161.248 g/mol

4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylpiperidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 771-99-3
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00006002
InChI Key UTBULQCHEUWJNV-UHFFFAOYSA-N
Synonym 4-phenyl-piperidine, piperidine, 4-phenyl, 4-phenyl piperidine, chembl20969, 4-phenypiperdine, 4-phenyl piperidin, pubchem8009, 4-phenylpiperidine
PubChem CID 69873
IUPAC Name 4-phenylpiperidine
SMILES C1CNCCC1C2=CC=CC=C2

Specifications

Melting Point 61°C to 64°C
Color Yellow to Brown
Quantity 1 g
Beilstein 124508
Solubility Information Soluble in water.
Formula Weight 161.24
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Phenylpiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 212-243-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only