missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcyclohexanone 98.0+%, TCI America™

Catalog No. p-7134882
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
P097525G 25 g
P0975250G 250 g
2 options

Catalog No. P097525G

Supplier: TCI America P097525G

Only null left

Chemical Identifiers

CAS 4894-75-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00001641
InChI Key YKAYMASDSHFOGI-UHFFFAOYSA-N
Synonym 4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
PubChem CID 78605
IUPAC Name 4-phenylcyclohexan-1-one
SMILES C1CC(=O)CCC1C2=CC=CC=C2

Specifications

Melting Point 78°C
Color White
Quantity 25 g
Formula Weight 174.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenylcyclohexanone