missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcyclohexanone, 98+%

Catalog No. p-7044886
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0861414 25 g
AAL0861422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0861414 Supplier Thermo Scientific Chemicals Supplier No. L0861414

CAS: 4894-75-1 | C12H14O | 174.243 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4894-75-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00001641
InChI Key YKAYMASDSHFOGI-UHFFFAOYSA-N
Synonym 4-phenylcyclohexanone, cyclohexanone, 4-phenyl, 4-phenyl-1-cyclohexanone, 4-phenyl cyclohexanone, 4-phenyl-cyclohexanone, 4-phenylcyclo-hexanone, acmc-1aoge, enamine_005496, 1-n-phenylpiperidin-4-one, 4-phenyl-cyclohexan-1-one
PubChem CID 78605
IUPAC Name 4-phenylcyclohexan-1-one
SMILES C1CC(=O)CCC1C2=CC=CC=C2

Specifications

Melting Point 78°C to 82°C
Boiling Point 158°C to 160°C (12 mmHg)
Flash Point >100°C (212°F)
Quantity 25 g
Beilstein 2045904
Formula Weight 174.24
Percent Purity ≥98%
Chemical Name or Material 4-Phenylcyclohexanone
EINECSNumber 225-517-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only