missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylcinnamic acid, 98%

Catalog No. AAA1264309
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1264309 10 g
AAA1264314 25 g
AAA1264318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1264309 Supplier Thermo Scientific Chemicals Supplier No. A1264309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13026-23-8 | C15H12O2 | 224.26 g/mol

4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylcinnamic acid is used as pharmaceutical intermediates and for chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 13026-23-8
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.26
MDL Number MFCD00014010
InChI Key DMJDEZUEYXVYNO-FLIBITNWSA-N
Synonym 4-phenylcinnamic acid, 3-1,1'-biphenyl-4-yl acrylic acid, p-phenylcinnamic acid, e-3-1,1'-biphenyl-4-yl acrylic acid, 2e-3-4-phenylphenyl prop-2-enoic acid, 2e-3-biphenyl-4-ylacrylic acid, 3-4-phenylphenyl prop-2-enoic acid, 2e-3-1,1'-biphenyl-4-yl prop-2-enoic acid, trans-4-phenylcinnamic acid, biphenylylacrylicacid
PubChem CID 5842785
IUPAC Name (E)-3-(4-phenylphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C/C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C to 228°C
Quantity 10 g
Beilstein 1955124
Solubility Information Slightly soluble in water.
Formula Weight 224.27
Percent Purity 98%
Chemical Name or Material 4-Phenylcinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-888-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only