missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, 97%

Catalog No. AC417360250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417360250 25 g Glass bottle
AC417361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417360250 Supplier Thermo Scientific Chemicals Supplier No. 417360250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1689-82-3 | C12H10N2O | 198.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 1689-82-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002330
InChI Key JTSBGMZPPPULTA-UHFFFAOYSA-N
Synonym C.I. 11800, 4-Hydroxyazobenzene, Solvent Yellow 7
PubChem CID 5354198
IUPAC Name 4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES O=C1C=CC(C=C1)=NNC1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 16, 96
Infrared Spectrum Authentic
Formula Weight 198.22
Percent Purity ≥96.0%
Packaging Glass bottle
Color Brown
Quantity 25 g
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Phenylazophenol
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only