missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenylazo)phenol 95.0+%, TCI America™
Supplier: TCI America P014925G
Specifications
| 4-(Phenylazo)phenol | |
| 230°C | |
| C12H10N2O | |
| 4-Hydroxyazobenzene, Solvent Yellow 7 | |
| O=C1C=CC(C=C1)=NNC1=CC=CC=C1 | |
| 198.23 | |
| 198.23 | |
| 25 g |
| 153°C | |
| 1689-82-3 | |
| MFCD00002330 | |
| JTSBGMZPPPULTA-UHFFFAOYSA-N | |
| 4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one | |
| 5354198 | |
| ≥95.0% (T) | |
| Crystal/Powder |
Chemical Identifiers
| 1689-82-3 | |
| 198.23 | |
| JTSBGMZPPPULTA-UHFFFAOYSA-N | |
| 5354198 | |
| O=C1C=CC(C=C1)=NNC1=CC=CC=C1 |
| C12H10N2O | |
| MFCD00002330 | |
| 4-Hydroxyazobenzene, Solvent Yellow 7 | |
| 4-(2-phenylhydrazin-1-ylidene)cyclohexa-2,5-dien-1-one |
Safety and Handling
EINECSNumber : (5)-3041
RTECSNumber : SM8300000
TSCA : Yes