missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%

Catalog No. p-183489
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122920250 25 g Glass bottle
AC122921000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122920250 Supplier Thermo Scientific Chemicals Supplier No. 122920250
Only null left

CAS: 83949-32-0 | C12H15NO2·C7H8O3S | 377.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83949-32-0
Molecular Formula C12H15NO2·C7H8O3S
Molecular Weight (g/mol) 377.45
MDL Number MFCD00044709
InChI Key NQLZTDKDXBKUGY-UHFFFAOYSA-N
Synonym 4-methylbenzene-1-sulfonic acid; 4-phenylpiperidine-4-carboxylic acid, 4-carboxy-4-phenylpiperidine tosylate, 4-phenylpiperidine-4-carboxylic acid compound with 4-methylbenzenesulfonic acid 1:1, 4-phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 4-phenyl-4-piperidinecarboxylic acid 4-methylbenzenesulfonate, 4-phenylpiperidine-4-carboxylic acid 4-toluenesulfonate, isonipecotic acid, 4-phenyl-; para-toluene sulfonate, 4-phenyl-4-carboxy piperidine tosylate, 4-carboxy-4-phenylpiperidine p-toluenesulfonate, 4-phenyl-piperidine-4-carboxylic acid tosylate
PubChem CID 2723657
IUPAC Name 4-methylbenzenesulfonic acid;4-phenylpiperidine-4-carboxylic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.C1CNCCC1(C2=CC=CC=C2)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 225°C to 229°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information (3% in boiling water) Clear colorless to almost colorless
Formula Weight 377.45
Percent Purity 98%
Chemical Name or Material 4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 281-391-7

RUO – Research Use Only