missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-butyn-2-one 97.0+%, TCI America™

Numéro de catalogue. p-7135084
Change view
Click to view available options
Quantity:
1 g
5 g
Numéro de catalogue. Quantity
P13361G 1 g
P13365G 5 g
2 options

Numéro de catalogue. P13361G

Supplier: TCI America P13361G

Il en reste null

Identifiants chimiques

CAS 1817-57-8
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00008776
InChI Key UPEUQDJSUFHFQP-UHFFFAOYSA-N
Synonym 4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
PubChem CID 74555
ChEBI CHEBI:51731
IUPAC Name 4-phenylbut-3-yn-2-one
SMILES CC(=O)C#CC1=CC=CC=C1

Spécifications

Color Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 144.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Phenyl-3-butyn-2-one
RTECSNumber ES0890000
TSCA No