missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-3-butyn-2-one, 98%, Thermo Scientific™

Catalog No. AC171330050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC171330050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171330050 Supplier Thermo Scientific Chemicals Supplier No. 171330050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1817-57-8
Density 0.9900g/mL
Boiling Point 75°C to 76°C (1.1 hPa)
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C10H8O
Refractive Index 1.5762
Linear Formula C6H5C≡CCOCH3
Quantity 5g
Beilstein 07,385
Specific Gravity 0.99
Synonym 4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
InChI Key UPEUQDJSUFHFQP-UHFFFAOYSA-N
SMILES CC(=O)C#CC1=CC=CC=C1
IUPAC Name 4-phenylbut-3-yn-2-one
Molecular Weight (g/mol) 144.17
PubChem CID 74555
ChEBI CHEBI:51731
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material 4-Phenyl-3-butyn-2-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 217-327-1

RUO – Research Use Only